Tisk…

Retinol all trans, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: all-trans-Retinol, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol, Vitamin A alcohol

95144-250MG 95144-1G
SIAL95144-250MGEA 5340 CZK
SIAL95144-250MG SIAL95144-1G
Retinol all trans, Sigma-Aldrich®
Retinol all trans
Vzorec: C₂₀H₃₀O
MW: 286,46 g/mol
MDL Number: MFCD00001552
CAS číslo: 68-26-8
EINECS: 200-683-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR