Tisk…

Triethyloxonium tetrafluoroborate stabilizovaný, Sigma-Aldrich®

Dodavatel: Merck


Stabilization: Stabilised with diethyl ether
M-Clarity™ quality level = MQ100

90520-100G 90520-25G
SIAL90520-100GEA 11300 CZK
SIAL90520-100G SIAL90520-25G
Triethyloxonium tetrafluoroborate stabilizovaný, Sigma-Aldrich®
Triethyloxonium tetrafluoroborate
Vzorec: C₆H₁₅BF₄O
MDL Number: MFCD00044423
CAS číslo: 368-39-8
EINECS: 206-705-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR