Tisk…

Terpineol, mixture of isomers, Sigma-Aldrich®

Dodavatel: Merck

86480-1L 86480-250ML 86480-4X25ML 86480-2.5L
SIAL86480-1LEA 5910 CZK
SIAL86480-1L SIAL86480-250ML SIAL86480-4X25ML SIAL86480-2.5L
Terpineol, mixture of isomers, Sigma-Aldrich®
Terpineol, mixture of isomers
Vzorec: C₁₀H₁₈O
MW: 154,25 g/mol
MDL Number: MFCD00166983
CAS číslo: 8000-41-7
EINECS: 232-268-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR