Tisk…

Pyridinium-p-toluenesulphonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Pyridin-1-ium 4-methylbenzenesulphonate

M-Clarity™ quality level = MQ200

82815-100G 82815-25G
SIAL82815-100GEA 16740 CZK
SIAL82815-100G SIAL82815-25G
Pyridinium-p-toluenesulphonate, Sigma-Aldrich®
Pyridinium-p-toluenesulphonate
Vzorec: C₁₂H₁₃NO₃S
MW: 251,31 g/mol
MDL Number: MFCD00013108
CAS číslo: 24057-28-1
EINECS: 246-002-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR