Tisk…

5-Nitro-2-furoyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Nitrofuran-2-carbonyl chloride

M-Clarity™ quality level = MQ100

683752-25G 683752-5G
SIAL683752-25GEA 22700 CZK
SIAL683752-25G SIAL683752-5G
5-Nitro-2-furoyl chloride, Sigma-Aldrich®
5-Nitro-2-furoyl chloride
Vzorec: C₅H₂ClNO₄
MW: 175,53 g/mol
MDL Number: MFCD00039564
CAS číslo: 25084-14-4
EINECS: 246-607-6
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR