Tisk…

3-Methyl-2-furoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Methylfuran-2-carboxylic acid

M-Clarity™ quality level = MQ100

667994-1G 667994-5G
SIAL667994-1GEA 3270 CZK
SIAL667994-1G SIAL667994-5G
3-Methyl-2-furoic acid, Sigma-Aldrich®
3-Methyl-2-furoic acid
Vzorec: C₆H₆O₃
MW: 126,11 g/mol
MDL Number: MFCD00014111
CAS číslo: 4412-96-8
EINECS: 224-571-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR