Tisk…

2-Acetamido-5-bromopyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Acetylamino-5-bromopyridine, N-(5-Bromopyridin-2-yl)acetamide

M-Clarity™ quality level = MQ100

646407-10G 646407-1G
SIAL646407-10GEA 0 CZK
SIAL646407-10G SIAL646407-1G
2-Acetamido-5-bromopyridine, Sigma-Aldrich®
2-Acetamido-5-bromopyridine
Vzorec: C₇H₇BrN₂O
MW: 215,05 g/mol
MDL Number: MFCD00468968
CAS číslo: 7169-97-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR