Tisk…

3-Fluoro-4-formylpyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Fluoro-4-pyridinecarbaldehyde, 3-Fluoroisonicotinaldehyde, 3-Fluoro-4-pyridinecarboxaldehyde

644315-1G 644315-5G
SIAL644315-1GEA 4600 CZK
SIAL644315-1G SIAL644315-5G
3-Fluoro-4-formylpyridine, Sigma-Aldrich®
3-Fluoro-4-formylpyridine
Vzorec: C₆H₄FNO
MW: 125,1 g/mol
MDL Number: MFCD06410686
CAS číslo: 40273-47-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR