Tisk…

Trifluoromethanesulphonamide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

638455-1G 638455-5G
SIAL638455-1GEA 3060 CZK
SIAL638455-1G SIAL638455-5G
Trifluoromethanesulphonamide, Sigma-Aldrich®
Trifluoromethanesulphonamide
Vzorec: CH₂F₃NO₂S
MDL Number: MFCD00068714
CAS číslo: 421-85-2
EINECS: 431-270-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR