Bismuth(III) trifluoromethanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Bismuth triflate, Bismuth(III) trifluoromethanesulphonate
M-Clarity™ quality level = MQ100
633305-25G
633305-5G
SIAL633305-25GEA
7930
CZK
SIAL633305-25G
SIAL633305-5G
Bismuth(III) trifluoromethanesulphonate, Sigma-Aldrich®
Bismuth(III) trifluoromethanesulphonate
Vzorec:
C₃BiF₉O₉S₃ MW: 656,19 g/mol |
MDL Number:
MFCD02093669 CAS číslo: 88189-03-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...