Tisk…

2-Bromo-3-formylpyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde

M-Clarity™ quality level = MQ100

632147-1G 632147-5G
SIAL632147-1GEA 2370 CZK
SIAL632147-1G SIAL632147-5G
2-Bromo-3-formylpyridine, Sigma-Aldrich®
2-Bromo-3-formylpyridine
Vzorec: C₆H₄BrNO
MW: 186,01 g/mol
MDL Number: MFCD04966945
CAS číslo: 128071-75-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR