Tisk…

Vinyl laurate, Sigma-Aldrich®

Dodavatel: Merck

61725-1L 61725-250ML
SIAL61725-1LEA 6990 CZK
SIAL61725-1L SIAL61725-250ML
Vinyl laurate, Sigma-Aldrich®
Vinyl laurate
Vzorec: C₁₄H₂₆O₂
MDL Number: MFCD00048421
CAS číslo: 2146-71-6
EINECS: 218-414-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR