Tisk…

Lithium-bis(trifluormethansulfonyl)imid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

544094-25G 544094-5G
SIAL544094-25GEA 5740 CZK
SIAL544094-25G SIAL544094-5G
Lithium-bis(trifluormethansulfonyl)imid, Sigma-Aldrich®
Lithium-bis(trifluormethansulfonyl)imid
Vzorec: C₂F₆LiNO₄S₂
MDL Number: MFCD00210017
CAS číslo: 90076-65-6
EINECS: 415-300-0
UN: 2923
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR