Tisk…

3-Formyl-6-methoxypyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

533068-1G 533068-25G 533068-5G
SIAL533068-1GEA 3740 CZK
SIAL533068-1G SIAL533068-25G SIAL533068-5G
3-Formyl-6-methoxypyridine, Sigma-Aldrich®
3-Formyl-6-methoxypyridine
Vzorec: C₇H₇NO₂
MDL Number: MFCD02683446
CAS číslo: 65873-72-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR