Tisk…

Diheptyl phthalate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

454818-10G 454818-50G
SIAL454818-10GEA 5000 CZK
SIAL454818-10G SIAL454818-50G
Diheptyl phthalate, Sigma-Aldrich®
Diheptyl phthalate
Vzorec: C₂₂H₃₄O₄
MDL Number: MFCD00043680
CAS číslo: 3648-21-3
EINECS: 222-885-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR