Dichloro(methyl)(phenyl)silane, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Methylphenyldichlorosilane
M-Clarity™ quality level = MQ100
440116-100ML
440116-500ML
SIAL440116-100MLEA
2540
CZK
SIAL440116-100ML
SIAL440116-500ML
Dichloro(methyl)(phenyl)silane, Sigma-Aldrich®
Dichloro(methyl)(phenyl)silane
Vzorec:
C₇H₈Cl₂Si |
MDL Number:
MFCD00000490 CAS číslo: 149-74-6 EINECS: 205-746-2 UN: 2437 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...