Tisk…

Brij® 98, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

436240-100G 436240-500G
SIAL436240-100GEA 1400 CZK
SIAL436240-100G SIAL436240-500G
Brij® 98, Sigma-Aldrich®
Brij® 98
Vzorec: C₅₈H₁₁₆O₂₁
Storage Temperature: Ambient
MDL Number: MFCD00074369
CAS číslo: 9004-98-2
EINECS: 500-016-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR