Tisk…

Lanthanum(III) trifluoromethanesulphonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Lanthanum(III) trifluoromethanesulphonate, Lanthanum trifluoromethanesulphonate, Lanthanum triflate

M-Clarity™ quality level = MQ100

430609-5G 430609-25G
SIAL430609-5GEA 2820 CZK
SIAL430609-5G SIAL430609-25G
Lanthanum(III) trifluoromethanesulphonate, Sigma-Aldrich®
Lanthanum(III) trifluoromethanesulphonate
Vzorec: C₃F₉LaO₉S₃
MW: 586.12
MDL Number: MFCD00077560
CAS číslo: 52093-26-2
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR