Tisk…

Erbium(III) trifluoromethanesulphonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

425672-25G 425672-5G
SIAL425672-25GEA 7280 CZK
SIAL425672-25G SIAL425672-5G
Erbium(III) trifluoromethanesulphonate, Sigma-Aldrich®
Erbium(III) trifluoromethanesulphonate
Vzorec: C₃ErF₉O₉S₃
MDL Number: MFCD00192126
CAS číslo: 139177-64-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR