1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
PYR14-TFSI, N-Butyl-N-methylpyrrolidinium bis((trifluoromethyl)sulphonyl)imide
38894-50G-F
38894-5G-F
SIAL38894-50G-FEA
17890
CZK
SIAL38894-50G-F
SIAL38894-5G-F
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide, Sigma-Aldrich®
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide
Vzorec:
C₁₁H₂₀F₆N₂O₄S₂ MW: 422,41 g/mol |
MDL Number:
MFCD07784447 CAS číslo: 223437-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...