Tisk…

Tantalum(V) ethanolate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tantalum(V) ethoxide

M-Clarity™ quality level = MQ100

339113-100G 339113-10G
SIAL339113-100GEA 38410 CZK
SIAL339113-100G SIAL339113-10G
Tantalum(V) ethanolate, Sigma-Aldrich®
Tantalum(V) ethanolate
Vzorec: C₁₀H₂₅O₅Ta
MDL Number: MFCD00049785
CAS číslo: 6074-84-6
EINECS: 228-010-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR