Magnesium trifluoromethanesulphonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Magnesium trifluoromethanesulphonate, Magnesium triflate
M-Clarity™ quality level = MQ100
337986-25G
337986-5G
SIAL337986-25GEA
3110
CZK
SIAL337986-25G
SIAL337986-5G
Magnesium trifluoromethanesulphonate, Sigma-Aldrich®
Magnesium trifluoromethanesulphonate
Vzorec:
C₂F₆MgO₆S₂ MW: 322.45 |
MDL Number:
MFCD00042601 CAS číslo: 60871-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...