Magnesium ethanolate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Magnesium ethoxide, Magnesium ethylate
M-Clarity™ quality level = MQ200
291846-100G
291846-500G
SIAL291846-100GEA
2080
CZK
SIAL291846-100G
SIAL291846-500G
Magnesium ethanolate, Sigma-Aldrich®
Magnesium ethanolate
Vzorec:
C₄H₁₀MgO₂ MW: 114,43 g/mol |
MDL Number:
MFCD00009060 CAS číslo: 2414-98-4 EINECS: 219-323-5 UN: 3205 ADR: 4.2,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...