Tisk…

Zinc trifluoromethanesulphonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Zinc(II) trifluoromethanesulphonate, Zinc triflate, Zinc trifluoromethanesulphonate

M-Clarity™ quality level = MQ200

290068-10G 290068-50G
SIAL290068-10GEA 3070 CZK
SIAL290068-10G SIAL290068-50G
Zinc trifluoromethanesulphonate, Sigma-Aldrich®
Zinc trifluoromethanesulphonate
Vzorec: C₂F₆O₆S₂Zn
MW: 363.53
MDL Number: MFCD00013229
CAS číslo: 54010-75-2
EINECS: 258-922-6
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR