Tisk…

Lithium trifluoromethanesulphonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

282669-100G 282669-25G
SIAL282669-100GEA 8750 CZK
SIAL282669-100G SIAL282669-25G
Lithium trifluoromethanesulphonate, Sigma-Aldrich®
Lithium trifluoromethanesulphonate
Vzorec: CF₃LiO₃S
MDL Number: MFCD00013227
CAS číslo: 33454-82-9
EINECS: 251-528-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR