Tisk…

Triphenylphosphine dibromide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

270946-25G 270946-5G
SIAL270946-25GEA 3900 CZK
SIAL270946-25G SIAL270946-5G
Triphenylphosphine dibromide, Sigma-Aldrich®
Triphenylphosphine dibromide
Vzorec: C₁₈H₁₅Br₂P
MDL Number: MFCD00000054
CAS číslo: 1034-39-5
EINECS: 213-855-1
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR