Tisk…

Aluminium triethanolate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Aluminumethoxide, Aluminum ethoxide, Aluminium triethylate

M-Clarity™ quality level = MQ200

235857-100G 235857-25G
SIAL235857-100GEA 11180 CZK
SIAL235857-100G SIAL235857-25G
Aluminium triethanolate, Sigma-Aldrich®
Aluminium triethanolate
Vzorec: C₆H₁₅AlO₃
MW: 162.16
MDL Number: MFCD00009658
CAS číslo: 555-75-9
EINECS: 209-105-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR