Pyridinium-p-toluenesulphonate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
232238-100G
232238-25G
232238-5G
SIAL232238-100GEA
5780
CZK
SIAL232238-100G
SIAL232238-25G
SIAL232238-5G
Pyridinium-p-toluenesulphonate, Sigma-Aldrich®
Pyridinium-p-toluenesulphonate
Vzorec:
C₁₂H₁₃NO₃S |
MDL Number:
MFCD00013108 CAS číslo: 24057-28-1 EINECS: 246-002-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...