Tris(triphenylphosphine)ruthenium(II) chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Dichlorotris(triphenylphosphine)ruthenium(II), Dichlorotris(triphenylphosphino)ruthenium (II)
M-Clarity™ quality level = MQ100
223662-1G
223662-25G
223662-5G
SIAL223662-1GEA
1870
CZK
SIAL223662-1G
SIAL223662-25G
SIAL223662-5G
Tris(triphenylphosphine)ruthenium(II) chloride, Sigma-Aldrich®
Tris(triphenylphosphine)ruthenium(II) chloride
Vzorec:
C₅₄H₄₅Cl₂P₃Ru MW: 958.85 |
MDL Number:
MFCD00013077 CAS číslo: 15529-49-4 EINECS: 239-569-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...