Tisk…

Ethyl-3-methylcrotonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,3-Dimethylacrylic acid ethyl ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-methylbut-2-enoate, 3-Methyl-2-butenoic acid ethyl ester, 3-Methylcrotonic acid ethyl ester, Ethyl 3-Methyl-2-butenoate

M-Clarity™ quality level = MQ100

194328-100G 194328-25G
SIAL194328-100GEA 10780 CZK
SIAL194328-100G SIAL194328-25G
Ethyl-3-methylcrotonate, Sigma-Aldrich®
Ethyl-3-methylcrotonate
Vzorec: C₇H₁₂O₂
MDL Number: MFCD00009187
CAS číslo: 638-10-8
EINECS: 211-319-1
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR