Retinol all trans, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
all-trans-Retinol, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-ol, Vitamin A alcohol
M-Clarity™ quality level = MQ100
17772-1G
17772-250MG
SIAL17772-1GEA
16000
CZK
SIAL17772-1G
SIAL17772-250MG
Retinol all trans, Sigma-Aldrich®
Retinol all trans
Vzorec:
C₂₀H₃₀O MW: 286,46 g/mol |
MDL Number:
MFCD00001552 CAS číslo: 68-26-8 EINECS: 200-683-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...