Tisk…

2-Picolyl chloride hydrochloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

162701-25G 162701-100G 162701-5G
SIAL162701-25GEA 4580 CZK
SIAL162701-25G SIAL162701-100G SIAL162701-5G
2-Picolyl chloride hydrochloride, Sigma-Aldrich®
2-Picolyl chloride hydrochloride
Vzorec: C₆H₇Cl₂N
MDL Number: MFCD00012811
CAS číslo: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR