Tisk…

2-Furoyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

149861-100G 149861-25G
SIAL149861-100GEA 2860 CZK
SIAL149861-100G SIAL149861-25G
2-Furoyl chloride, Sigma-Aldrich®
2-Furoyl chloride
Vzorec: C₅H₃ClO₂
MW: 130,53 g/mol
MDL Number: MFCD00003228
CAS číslo: 527-69-5
EINECS: 208-422-9
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR