2-Bromoterephthalic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
115274-25G
115274-5G
SIAL115274-25GEA
9810
CZK
SIAL115274-25G
SIAL115274-5G
2-Bromoterephthalic acid, Sigma-Aldrich®
2-Bromoterephthalic acid
Vzorec:
C₈H₅BrO₄ |
MDL Number:
MFCD00002403 CAS číslo: 586-35-6 EINECS: 209-572-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...