Methyltrioctylammonium bis(trifluoromethylsulphonyl)imide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N-Methyl-N,N-dioctyloctan-1-aminium bis((trifluoromethyl)sulphonyl)amide
MQ100
00797-50G-F
00797-5G-F
SIAL00797-50G-FEA
14330
CZK
SIAL00797-50G-F
SIAL00797-5G-F
Methyltrioctylammonium bis(trifluoromethylsulphonyl)imide, Sigma-Aldrich®
Methyltrioctylammonium bis(trifluoromethylsulphonyl)imide
Vzorec:
C₂₇H₅₄F₆N₂O₄S₂ MW: 648.86 |
MDL Number:
MFCD08275364 CAS číslo: 375395-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...