Ethyl vinyl ether stabilizovaný, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Stabilization:
Stabilised with potassium hydroxide
M-Clarity™ quality level = MQ200
8.01391.0100
8.01391.1000
8.01391.0100EA
884
CZK
8.01391.0100
8.01391.1000
Ethyl vinyl ether stabilizovaný, Sigma-Aldrich®
Ethyl vinyl ether
Vzorec:
C₄H₈O Storage Temperature: Refrigerator |
MDL Number:
MFCD00009248 CAS číslo: 109-92-2 EINECS: 203-718-4 UN: 1302 ADR: 3,I |
Výsledky specifikačního testu
Solubility | 8.3 g/l (15 °C) |
Melting Point | -115 °C |
Molar Mass | 72.11 g/mol |
Boiling Point | 36 °C (1013 hPa) |
Vapor Pressure | 560 hPa (20 °C) |
Flash Point | -45 °C |
Refractive Index | 1.3757 (20 °C, 589 nm) |
Explosion Limit | 1.3 - 12.0 %(V) |
Density | 0.75 g/cm3 (20 °C) |
Ignition Point | 178 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...