Tisk…

Dysprosium(III) trifluoromethanesulphonate 98%

Dodavatel: Thermo Scientific
Varování

L20252.06 L20252.14
L20252.06EA 2300 CZK
L20252.06 L20252.14
Dysprosium(III) trifluoromethanesulphonate 98%
Dysprosium(III) trifluoromethanesulphonate

Purity: 98% CAS No.: 139177-62-1 Molecular Formula: C3DyF9O9S3 Formula Weight: 609.69 MDL No.: MFCD00209583

Vzorec: C₃DyF₉O₉S₃
MW: 609,71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209583
CAS číslo: 139177-62-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR