Tisk…

1-Boc-4-hydroxypiperidine ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L19275.06 L19275.14
L19275.06EA 831 CZK
L19275.06 L19275.14
1-Boc-4-hydroxypiperidine ≥98%
1-Boc-4-hydroxypiperidine

Purity: 98% CAS No.: 109384-19-2 Molecular Formula: C10H19NO3 Formula Weight: 201.27 Melting Point: 62-64° Beilstein Reference: 7202094

Vzorec: C₁₀H₁₉NO₃
Bod tání: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD01075174
CAS číslo: 109384-19-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR