Tisk…

3-Chlor-1,2-benzisothiazol ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

L17791.06 L17791.14
L17791.06EA 2210 CZK
L17791.06 L17791.14
3-Chlor-1,2-benzisothiazol ≥97%
3-Chlor-1,2-benzisothiazol

Purity: 99% CAS No.: 7716-66-7 Molecular Formula: C7H4ClNS Formula Weight: 169.63 Melting Point: 37-44° Boiling Point: 80-86°/0.75mm MDL No.: MFCD00673254

Vzorec: C₇H₄ClNS
Bod varu: 80…86 °C (0,75 mmHg)
Bod tání: 37…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00673254
CAS číslo: 7716-66-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR