Tisk…

4,5-Dibromo-2-furoic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4,5-Dibromofuran-2-carboxylic acid

L15517.06 L15517.14
L15517.06EA 1970 CZK
L15517.06 L15517.14
4,5-Dibromo-2-furoic acid ≥97%
4,5-Dibromo-2-furoic acid

Purity: 97% CAS No.: 2434-03-9 Molecular Formula: C5H2Br2O3 Formula Weight: 269.88 Melting Point: 169-172° Beilstein Reference: 129312

Vzorec: C₅H₂Br₂O₃
MW: 269.88
Bod tání: 169…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00092311
CAS číslo: 2434-03-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR