Methyl-3-bromopropionate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L14127.14
L14127.22
L14127.14EA
2360
CZK
L14127.14
L14127.22
Methyl-3-bromopropionate ≥97%
Methyl-3-bromopropionate
Purity: 97%
CAS No.: 3395-91-3
Molecular Formula: Br(CH2)2CO2CH3
Formula Weight: 167.01
Flash Point: 75°(167°F)
Refractive Index: 1.4580
Boiling Point: 58-60°/12mm
Beilstein Reference: 1742949
MDL No.: MFCD00000250
Vzorec:
C₄H₇BrO₂ Bod varu: 58…60 °C (12 mmHg) Hustota: 1,530 Bod vzplanutí: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000250 CAS číslo: 3395-91-3 EINECS: 222-247-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...