L-α-Aminoadipic acid 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
L-2-Aminoadipic acid, (S)-2-Aminoadipic Acid
L13924.03
L13924.06
L13924.03EA
2820
CZK
L13924.03
L13924.06
L-α-Aminoadipic acid 98%
L-α-Aminoadipic acid
Purity: 98%
CAS No.: 1118-90-7
Molecular Formula: C6H11NO4
Formula Weight: 161.16
Melting Point: ca 206° dec.
Beilstein Reference: 1724348
Vzorec:
C₆H₁₁NO₄ MW: 161.16 Bod tání: ∼ 206 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00002636 CAS číslo: 1118-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...