Tisk…

Tantalum(V) ethanolate ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L10288.06 L10288.14
L10288.06EA 3350 CZK
L10288.06 L10288.14
Tantalum(V) ethanolate ≥99%
Tantalum(V) ethanolate

Purity: 99+% CAS No.: 6074-84-6 Molecular Formula: C10H25O5Ta Formula Weight: 406.25 Flash Point: 30°(86°F) Refractive Index: 1.4880 Melting Point: 18-19° Boiling Point: 145°/0.1mm Beilstein Reference: 3678999 UN No.: 2924 MDL No.: MFCD00049785

Vzorec: C₁₀H₂₅O₅Ta
Bod varu: 145 °C (0,1 mmHg)
Bod tání: 18…19 °C
Hustota: 1,566
Bod vzplanutí: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00049785
CAS číslo: 6074-84-6
EINECS: 228-010-2
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR