m-Toluic hydrazide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
m-Toluic acid hydrazide, 3-Methylbenzoic hydrazide
L01043.06
L01043.14
L01043.06EA
1240
CZK
L01043.06
L01043.14
m-Toluic hydrazide ≥97%
m-Toluic hydrazide
Purity: 97%
CAS No.: 13050-47-0
Molecular Formula: C8H10N2O
Formula Weight: 150.18
Melting Point: 94-98°
Beilstein Reference: 2045125
Vzorec:
C₈H₁₀N₂O MW: 150.18 Bod tání: 94…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014760 CAS číslo: 13050-47-0 EINECS: 235-926-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...