5-Fluoro-2-nitrobenzonitrile ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H32756.03
H32756.06
H32756.03EA
1810
CZK
H32756.03
H32756.06
5-Fluoro-2-nitrobenzonitrile ≥97%
5-Fluoro-2-nitrobenzonitrile
Purity: 97%
CAS No.: 50594-78-0
Molecular Formula: C7H3FN2O2
Formula Weight: 166.11
Melting Point: 101-105°
UN No.: 3439
MDL No.: MFCD01632196
Vzorec:
C₇H₃FN₂O₂ Bod tání: 101…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632196 CAS číslo: 50594-78-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...