Tisk…

5-Bromo-2-isopropoxypyridine ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H32564.03 H32564.06
H32564.03EA 2980 CZK
H32564.03 H32564.06
5-Bromo-2-isopropoxypyridine ≥97%
5-Bromo-2-isopropoxypyridine

Purity: 97% CAS No.: 870521-31-6 Molecular Formula: C8H10BrNO Formula Weight: 216.08 Refractive Index: 1.5305

Vzorec: C₈H₁₀BrNO
MW: 216.08
Storage Temperature: Ambient
MDL Number: MFCD07368869
CAS číslo: 870521-31-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR