4-Fluoro-3-nitrobenzonitrile ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H32391.03
H32391.06
H32391.03EA
1070
CZK
H32391.03
H32391.06
4-Fluoro-3-nitrobenzonitrile ≥97%
4-Fluoro-3-nitrobenzonitrile
Purity: 97%
CAS No.: 1009-35-4
Molecular Formula: C7H3FN2O2
Formula Weight: 166.11
Physical Form: Solid
Appearance: Cream to orange
Melting Point: 90-94°
UN No.: 3439
MDL No.: MFCD01632197
Vzorec:
C₇H₃FN₂O₂ Bod tání: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632197 CAS číslo: 1009-35-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...