Methyl-3-bromo-4-hydroxybenzoate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Methyl 3-bromo-4-hydroxybenzoate
H30222.09
H30222.18
H30222.09EA
1570
CZK
H30222.09
H30222.18
Methyl-3-bromo-4-hydroxybenzoate ≥98%
Methyl-3-bromo-4-hydroxybenzoate
Purity: 98%
CAS No.: 29415-97-2
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Melting Point: 108-110°
MDL No.: MFCD06203850
Vzorec:
C₈H₇BrO₃ MW: 231,05 g/mol Bod tání: 108…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD06203850 CAS číslo: 29415-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...