Tisk…

1-Butyl-4-methylpyridiniumhexafluorophosphate ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Butyl-4-methylpyridinium Hexafluorophosphate

H27355.06 H27355.18
H27355.06EA 2900 CZK
H27355.06 H27355.18
1-Butyl-4-methylpyridiniumhexafluorophosphate ≥99%
1-Butyl-4-methylpyridiniumhexafluorophosphate

Purity: 99% CAS No.: 401788-99-6 Molecular Formula: C10H16F6NP Formula Weight: 295.22 Melting Point: 45° MDL No.: MFCD03095438

Vzorec: C₁₀H₁₆F₆NP
MW: 295,21 g/mol
Bod tání: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD03095438
CAS číslo: 401788-99-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR