4-Ethynyltoluene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B24812.06
B24812.14
B24812.06EA
2260
CZK
B24812.06
B24812.14
4-Ethynyltoluene ≥98%
4-Ethynyltoluene
Purity: 98%
CAS No.: 766-97-2
Molecular Formula: C9H8
Formula Weight: 116.16
Flash Point: 49°(120°F)
Refractive Index: 1.5470
Odour: Pungent
Melting Point: ca 21°
Boiling Point: 168-170°
Beilstein Reference: 969653
UN No.: 3295
MDL No.: MFCD00008571
Vzorec:
C₉H₈ Bod varu: 168…170 °C Bod tání: ∼ 21 °C Hustota: 0,916 Bod vzplanutí: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008571 CAS číslo: 766-97-2 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...